Can be written on using a pen, pencil, and/or a permanent marker without smearing. Can be adhered, removed, and reapplied without leaving behind the adhesive residue.
- Oil-resistant
- Acid-resistant
- Water-resistant
- Withstands temperatures ranging from −23 to 121 °C
Common in laboratories for color-coding or labeling.