Would you like to log in again?
DYCLEAN CONC CLEANING SOLUTION 1 US GAL
Supplier: DYCEM
Dyclean Conc Cleaning Solution 1 Us Gal^ Dyclean is an all-purpose, water soluble concentrated cleaning solution for regular cleaning in most applications outside the cleanroom environment, Use diluted 1:20 with water for regular maintenance, It can be used for heavy duty cleaning as needed
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DYCLEAN CONC CLEANING SOLUTION 1 US GAL 76376-590 CZCLDCGUS Each (1US Gal) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty