Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ActivArmr® 42-474 Series Heat Resistant Gloves, Ansell

Supplier: Ansell

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ActivArmr®
288352 288353 209337
76376-598CS 1789.86 USD InStock
76376-598 76376-600 76376-602
ActivArmr® 42-474 Series Heat Resistant Gloves, Ansell
Gloves
Superior heat resistance with complete protection for the hand and wrist.

  • Nonwoven felt insulation and a full-length heat barrier designed for intermittent handling of dry objects up to 200 °C (400 °F)
  • Two-piece construction eliminates out seams for better finger comfort, dexterity, and flexibility
  • Bonded nitrile coating protects against snags, punctures, abrasions, and cuts
  • Provides excellent grip when handling dry or oily surfaces
  • Contains no asbestos
  • Compliant to FDA food handling requirements

For the best protection and comfort in moderate heat applications, ActivArmr® 42-474 gloves are the answer. Their nonwoven felt insulation and full-length heat barrier protect the worker, while two-piece construction eliminates out seams to provide better finger comfort, dexterity, and flexibility. These gloves feature an exclusive bonded nitrile coating to help protect against snags, punctures, abrasions, and cuts, while also enhancing dry grip. ActivArmr® 42-474 gloves wear an average of four times longer than 24 ounce cotton hot mill gloves. These gloves were designed to handle intermittent handling of hot cores, castings, and controls up to 400 °F, not as a replacement for silicate gloves.

Certifications: ANSI A2 Cut, EN388:2016 (2241B), EN388:2003 (2241), EN407 (X2XXXX), EN420:2003 + A1:2009, Category III, REACH Compliant.

Packaging: Gloves are packaged 12 pairs per bag, 6 bags per carton.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty