Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ECHO Revolve 4, Hybrid Fluorescence Microscope (Upright/Inverted), Discover Echo
Supplier: ECHO
Revolve
PHAS0001 JCND2005 SCND1001 CUBE4005 PHAS0000 CUBE4004 RVSF1000 CUBE4002 CUBE4009 CUBE4011 CUBE4006 CUBE4008 CUBE4001 JCND2003 JCND2004 CUBE4003 CUBE4007
76490-302EA 31619.55 USD InStock
76490-302 76376-720 76490-316 76376-730 76490-310 76376-722 76376-732 76376-724 76376-734 76490-308 76376-726 76299-602 76299-612 76376-728 76299-610 76490-290 76376-718
ECHO Revolve 4, Hybrid Fluorescence Microscope (Upright/Inverted), Discover Echo
Microscopes
This 'all-in-one' hybrid microscope can be used for upright and inverted observation methods.
  • 5 MP monochrome camera (upgrade from 3.2 MP) - this increases field of view and resolution for fluorescence imaging
  • 5 MP CMOS color camera (brightfield)
  • Intelligent objective nosepiece - this allows the software interface to automatically recognise the objective being used
  • Illumination technique: Brightfield and Epi-fluorescence (phase contrast available as an option)
  • Ultra-bright LEDs with 50000-hour lifespan
  • Touch screen functions drive cameras and illumination, replacing manual turrets and levers
  • Traditional eyepieces are replaced by an iPad Pro® tablet for easier viewing (retina display)
  • Custom software App simplifies the workflow of taking images and seamlessly integrates with other cloud sharing Apps
  • Transmitted light, phase contrast, darkfield, polarized light and fluorescence in both upright and inverted configruations
The upright observation method is ideal for viewing glass slides. Useful for water dipping applications.
The inverted observation method is suitable for viewing live samples in fluidic chambers such as dishes and well plates. It also allows for easier manipulation of samples. Ideal for long working distance applications.
Accessories information: Z stack software module (optional) - automated acquisition of Z stacks.
Digital haze reduction software (optional).
Ordering information: To be completed with objectives, filters and light cubes, which are sold separately.
Delivery information: Supplied with an iPad® interface, motorised epi-fluorescence turret, brightfield and fluorescence cameras, power supply, precision X/Y stage, microscope cover, dark box, stage clip and lens cloth.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty