Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Cubis® II Advanced Premium Precision Laboratory Balances, Automatic, Standard Version, Sartorius
Supplier: Sartorius
Cubis® II
MCA623P-2S00-R MCA1203S-2S00-A MCA2203S-2S00-A MCA623P-2S00-A MCA623P-2S00-E MCA5203P-2S00-I MCA2203SR-S00 MCA1203S-2S00-U MCA3203S-2S00-I MCA5203S-2S00-R MCA2203P-2S00-U MCA5203SE-S00 MCA1203S-2S00-A MCA3203S-2S00-R MCA623SE-S00-E MCA3203SR-S00-R MCA3203S-2S00-A MCA5203P-2S00-U MCA3203S-2S00-I MCA1203S-2S00-R MCA5203P-2S00-E MCA2203P-2S00-A MCA623P-2S00-A MCA323S-2S00-A MCA5203P-2S00-A MCA2203SE-S00 MCA2203SR-S00-R MCA2203P-2S00-U MCA2203S-2S00-U MCA623SR-S00 MCA5203SE-S00-E MCA2203S-2S00-A MCA2203P-2S00-E MCA1203S-2S00-R MCA2203P-2S00-A MCA1203S-2S00-I MCA5203S-2S00-A MCA623P-2S00-I MCA5203S-2S00-U MCA5203P-2S00-A MCA1203S-2S00-U MCA3203S-2S00-A MCA3203S-2S00-U MCA3203S-2S00-E MCA323S-2S00-I MCA5203S-2S00-E MCA2203S-2S00-U MCA2203S-2S00-R MCA2203P-2S00-I MCA5203S-2S00-A MCA323S-2S00-I MCA323S-2S00-E MCA2203P-2S00-R MCA623S-2S00-E MCA2203S-2S00-A MCA5203P-2S00-R MCA5203S-2S00-I MCA5203P-2S00-U MCA3203SE-S00-E MCA5203SR-S00-R MCA323S-2S00-U MCA623S-2S00-R MCA623S-2S00-I MCA1203SE-S00 MCA3203S-2S00-R MCA5203S-2S00-U MCA5203S-2S00-R MCA623S-2S00-A MCA3203S-2S00-U MCA2203P-2S00-R MCA323S-2S00-R MCA623S-2S00-R MCA1203S-2S00-I MCA2203S-2S00-I MCA1203SE-S00-E MCA1203SR-S00 MCA3203SR-S00 MCA5203P-2S00-I MCA5203P-2S00-A MCA623SE-S00 MCA3203SE-S00 MCA2203S-2S00-I MCA2203P-2S00-I MCA1203SR-S00 MCA623S-2S00-U MCA5203P-2S00-R MCA2203SE-S00-E MCA3203S-2S00-A MCA623S-2S00-A MCA323S-2S00-E MCA623S-2S00-U MCA1203S-2S00-E MCA1203S-2S00-A MCA623P-2S00-E MCA623S-2S00-I MCA2203P-2S00-A MCA5203SR-S00 MCA623P-2S00-I MCA623P-2S00-U MCA5203S-2S00-I MCA2203S-2S00-R MCA323S-2S00-U MCA2203S-2S00-E MCA623P-2S00-R MCA1203S-2S00-E MCA5203S-2S00-A MCA623S-2S00-E MCA623SR-S00 MCA323S-2S00-R MCA623P-2S00-U
76377-132EA 12557.11 USD InStock
CA76334-018 CA76334-022 76377-132 CA76334-020 CA76334-142 CA76334-026 76377-094 CA76334-024 76377-092 76377-136 76377-256 76376-952 76376-954 CA76334-028 CA76334-034 76377-140 CA76334-032 CA76334-038 CA76334-036 76377-304 76376-948 76377-026 CA76334-030 76377-024 76376-942 CA76334-044 CA76377-164 CA76334-042 CA76377-288 CA76334-048 CA76334-046 CA76334-040 CA76377-162 76377-310 76377-044 76377-286 CA76334-056 76377-164 76377-162 CA76334-054 CA76334-058 76377-324 76377-048 CA76333-810 CA76334-052 76377-168 76377-288 76377-200 CA76334-050 76376-920 76376-922 76377-330 CA76334-066 76377-296 CA76334-064 CA76333-928 76377-292 76377-170 CA76334-068 76377-290 76377-216 76376-912 CA76333-804 CA76333-806 76377-214 CA76334-062 76377-058 CA76377-062 CA76334-060 76377-218 76376-910 CA76334-228 CA76334-078 CA76334-232 76377-064 CA76334-230 CA76334-076 76377-062 76377-182 CA76334-236 CA76334-234 CA76334-070 76377-104 76377-102 CA76334-074 CA76333-912 CA76334-072 76377-108 76377-190 CA76334-238 76377-110 CA76334-088 76377-076 CA76334-086 76377-194 76377-192 CA76334-080 76377-236 CA76334-084 76377-234 CA76334-082 76377-078 76376-972 76377-240 76377-128 CA76334-092 76377-248 CA76334-090 76377-244 CA76334-094
Cubis® II Advanced Premium Precision Laboratory Balances, Automatic, Standard Version, Sartorius
Balances and Scales
These MCA series advanced user interface laboratory balances have fully customizable hardware, software, and connectivity including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QApp packages for optional software extension.
  • Quick and easy, motorized, auto-levelling function, integrated climate sensors, individual sample holders
  • Error-free operation with individual QApp workflows and full end-to-end data integrity
  • Outstanding service support integrated status center, integrated service functions preventive maintenance on accredited standards
  • User Management provides complete access control and configured in accordance with password policy
  • Gesture control allows user to open and close the automatic draft shield, start the ionizer, or send a document
  • Maximum capacity between 320 and 14200 g scale interval with a range of 1 to 100 mg
  • Easy to use manual glass draft shield
  • Removable flat stainless-steel draft shield frame
Climate monitoring ensures that temperature, pressure, and humidity do not negatively impact results. Status center notifications show all information about balance and environmental conditions like calibration, leveling, temperature, humidity, air pressure and service, centralized on a dashboard. Remove balance stabilization frustrations caused by electrostatic effects with the built-in ionizing technology. It provides modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Audit trail and alibi memory with a tamper-protected time-stamped electronic logfile, that allows reconstruction of events related to the creation, modification, and or deletion of records and the system automatically stores weighing data in a ring buffer. The data can be displayed, filtered, and exported in an easy way. It can automatically execute time-controlled actions for backup. The system is able to upload data on a file share or export the data to other systems. Safe data transfer saves all files, together with a calculated MD5 checksum which is stored in the audit trail files. An electronic record should be protected against any manipulation. Built-in ionizer for electrostatic elimination- electrostatic charges on samples and vessels are removed effectively. For all models up to a maximum capacity of 8,2 kg.
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance for all weighing tasks. The Cubis® II range of premium laboratory balances with a maximum load between 2,1 and 70 kg provides the ideal model for every application. It supports automatic time synchronization through Network Time Protocol (NTP). An electronic signature can be used to sign the final report for weighing processes.
Display: Large high end 7" color touch TFT display in 16:9 format with new user interface; Software: Factory installed basic set of essential weighing applications (license free) and packages with special weighing applications and function extensions (license required); Operation: Activated by touch key, touch-free using IR sensor (draft shield M) or gesture sensor (optional), learning capability.
Draft shield: A- motorized draft shield; E - small glass draft shield; I - motorized draft shield + ionizer; R - stainless steel draft shield frame; U - manual glass draft shield.
Certifications: The Cubis® II balances ensure safe and accurate data storage and supports all requirements for FDA directive 21 CFR Part 11 compliance and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the user's workflows. Choice of QApp package: Pharma; advanced applications: utilities; Connectivity or the all inclusive QApps package includes all of these four packages. (denoted by QP99).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty