Would you like to log in again?
Cubis® II Advanced Premium Semi Micro Balances, MCA Series, Standard Versions, Sartorius
Supplier: Sartorius
Cubis® II
MCA225P-2S00-U MCA125S2S00I MCA125S-2S00-I MCA125P-2S00-A MCA125S2S00A MCA125S-2S00-A MCA225S2S00I MCA225P2S00I MCA125S2S00U MCA225SI-S00 MCA225S2S00A MCA225S-2S00-I MCA225P2S00U MCA125P2S00A MCA225P-2S00-I MCA125S-2S00-U MCA125SI-S00 MCA225P2S00A MCA125SI-S00 MCA225S-2S00-U MCA125P2S00U MCA125P-2S00-U MCA225SU-S00 MCA125P2S00I MCA225S-2S00-A MCA225S2S00U MCA225SI-S00 MCA225P-2S00-A MCA125P-2S00-I MCA225SU-S00
76377-352EA 25551.96 USD InStock
CA76334-000 76377-352 76377-274 76377-054 76377-252 76377-350 76377-174 CA76377-344 CA76334-004 CA76334-002 76377-316 76377-238 76377-336 76377-334 CA76333-964 CA76333-982 CA76333-960 CA76333-962 CA76333-984 CA76333-980 76377-340 CA76377-352 CA76377-336 76377-206 CA76333-958 76377-302 CA76333-998 76377-344 CA76333-978 76377-320
Cubis® II Advanced Premium Semi Micro Balances, MCA Series, Standard Versions, Sartorius
Balances and Scales
These Cubis® II laboratory balances are ideal for high-performance portfolio of both lab weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data intergrity, reliable support
  • Automatic glass draft shield with or without integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
  • Removable flat stainless steel draft shield frame
  • Large 7" color touch display including touch, scroll, swipe functionality
  • Diverse QApp packages for optional software extension
  • Automatic, motorized leveling, gesture control, Climate monitoring, built-in ionizer for electrostatic elimination
These balances offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Different draft sheilds are
A - Automatic, glass motorized draft shield with learning capability for user-friendly operation and easy customization to the changing requirements of different applications
I - Identical to the A draft shield, but also includes an integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
U - Manual glass analytical draft shield chamber, with smooth-action doors that open wide and provide unimpeded access to the weighing chamber without interfering braces.
Certifications: All requirements for FDA directive 21 CFR part 11 and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customise the balance for the users’ workflows. Choice of QApp Package: Pharma; Advanced Applications: Utilities; Connectivity or the all inclusive QApps package includes all of these four packages. (denoted by QP99).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty