Would you like to log in again?
TransPorter® Transport Swab, Amies Gel
Supplier: Hardy Diagnostics
TransPorter®
4142BX 4190BX 4191BX 4124BX 4192BX 4125BX 4140CQBX 4136BX
76377-440BX 117.93 USD InStock
76377-440 76377-450 76377-460 10790-373 76377-458 76377-454 76377-442 76377-452
TransPorter® Transport Swab, Amies Gel
Swabs Transport Swabs
Amies media is stuart's original media formula modified by replacing the glycerophosphate in the original formula for an inorganic phosphate buffer.
  • Hardy diagnostics offers amies medium in two formats
  • Unique tube shape preserves gel media
  • Maximum gel for anaerobe preservation
  • Available with or without charcoal
The metabolism of glycerophosphate by coliform organisms and other gram negative rods, in stuart's original formulation, resulted in proliferation of these organisms from throat, wound and fecal specimens. Amies also found that NaCl at 0.3% weight/volume was optimal for the preservation of Neisseria gonorrhoeae. Calcium and magnesium salts were added in the belief that these ions were of importance in controlling the permeability of the bacterial cells and contributed to their survival.
Liquid amies for aerobic culture amies agar gel (with and without charcoal) for aerobic and anaerobic culture. Unique venturi hourglass tube design prevents disintegration or breakdown of gel column during specimen transport and eliminates harmful air bubbles.
5 ml gel column affords maximum protection and improves sample viability. The presence of charcoal in the medium neutralizes toxins and inhibitory substances. Amies formula with charcoal has been shown to improve the recovery of Neisseria gonorrhoeae and is often recommended for Bordetella pertussis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty