Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Transporter, Liquid Stuart, Healthlink
Supplier: Hardy Diagnostics
4113BX 4029BX 4019BX 4111BX
76377-438BX 163.15 USD InStock
76377-438 76334-632 76377-448 76377-446
Transporter, Liquid Stuart, Healthlink
Swabs Transport Swabs
Stuart media, invented by Dr. R.D Stuart, was the first media for transporting clinical swab samples.
  • Liquid stuart medium bacteriology swab collection system
  • For rapid strep A antigen tests and aerobic culture
  • Safe, no glass ampoules to break, no seal or barriers to puncture or dry out swabs
  • No dry swabs - swabs are in direct contact with fluid-filled sponge, which acts as a moisture reservoir for the 1 ml of liquid transport media that is designed to stay in place during use
  • Multiple swab sizes and shaft material
Liquid Stuart swab collection systems are available in various sizes for multiple applications. Regular size swabs are typically used for throat, vaginal, wound, and skin samples. Minitip size swabs are typically used for pediatric sampling, ear, nose, throat, eye, and male urethral swab sampling. Swabs are also available with different shaft material depending on the application and preference of the user.
Hardy offers stuart medium in two formats, liquid stuart for rapid strep A antigen tests and aerobic culture and stuart agar gel for aerobic culture.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty