Would you like to log in again?
CRITERION™ LB Broth, Miller, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C9271 C9272
76446-886EA 111.48 USD InStock
76446-886 76377-544
CRITERION™ LB Broth, Miller, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ LB Broth, Miller is recommended for the cultivation and maintenance of Escherichia coli used in molecular biology procedures.
  • Wide mouth opening reduces dust formation by allowing the easy entry of a scoop
  • Stackable containers for efficient storage
  • Increased solubility dissolves quickly
  • Custom and bulk products available to your specifications
  • Complete technical/instruction sheets available in HUGO software
  • Unparalleled performance tested for optimal growth
Dehydrated culture media is a raw material not intended for use in the diagnosis of human disease. For implementation, this product requires additional processing and supplementation of ingredients before use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty