Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Fecal Swab Flocked Swab with Cary-Blair Medium, Copan Diagnostics
Supplier: COPAN Diagnostics
4C024S 4C028S
76377-560CS 1082.07 USD InStock
76377-560 76377-558
Fecal Swab Flocked Swab with Cary-Blair Medium, Copan Diagnostics
Swabs Transport Swabs
Patented sample collection and preservation system for enteric bacteria.
  • Smaller sample quantity, eliminating the need to vent the container, preventing messy accidents during processing
  • Compact alternative for space efficient transportation compared with traditional bulky fecal containers
  • Stool samples are easily processed on automated specimen processors, and automatic pipettors minimizing manual handling and mess and maximizing investment in automation
FecalSwab™ is a collection and transport system which is FDA cleared for rectal swab and fecal specimens for enteric pathogen recovery using traditional bacteriology culture.
The system is also compatible with enteric molecular assays for bacteria, viruses, and parasites where package inserts indicate Cary-Blair systems for sample collection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty