Would you like to log in again?
MicroOne® Pocket Mops, Vileda Professional
Supplier: Vileda Professional
Micro One®
163820 163818 163808 163810
76378-086CS 271.5 USD InStock
76378-086 76378-092 76378-090 76378-088
MicroOne® Pocket Mops, Vileda Professional
Mops
Hygiene and performance without compromise.
  • Microfiber cleaning performance in a disposable mop
  • Fiber is split into 32 segments vs. traditional 16 split, for improved cleaning results
  • Low friction and fast, stubborn dirt removal with optimized pocket design
  • Greater than 99.99% bacteria removal (including C-Diff, third party independently proven)
  • Additional inner layer increases absorbency and floor coverage by 30%
  • Fits CombiSpeed™ Pro frame in addition to other common industry frames
The pocket mop system is ideal for bucket and pre-prepared methods. The lightweight handle and frame used within the pocket mop system provides easy and effective cleaning with improved ergonomics.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty