Would you like to log in again?
PIG® Grippy® Adhesive-Backed Entrance Mats, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG® Grippy®
GRP926X10-GY GRP913X25-GY GRP923X25-GY GRP916X10-GY
76378-328RL 138.24 USD InStock
76378-328 76378-326 76378-324 76378-322
PIG® Grippy® Adhesive-Backed Entrance Mats, New Pig
Spill Control Materials
Keep floors safe and capture dirt and liquid at your door with these adhesive-backed mats. The carpeted absorbent runner comes in multiple textures.
  • P53™ adhesive backing delivers the ideal balance of stick-and-release performance
  • Sticks to most commercial floor surfaces including concrete, VCT, linoleum, ceramic tile, quarry tile and laminate flooring
  • Quick-dry, colorfast fibers resist staining and fading; made from 100% postconsumer recycled content
  • No rippled edges to obstruct doors or create tripping hazards; ADA compliant for wheelchairs and walkers
  • Commercial-grade durability lets you keep mat in place for three to six months depending on your conditions
  • Long-lasting carpet surface resists crushing to extend performance lif
  • Clean your floors with mats in place to save time and avoid the labor of dragging heavy mats around
  • EZCut™ design lets you roll out continuous walk-off coverage with no gaps, ripples or overlaps that cause slips and trips
  • Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents
Adhesive-backed mat stays exactly where you put it with no bunching, shifting, rippling, curling or flipping over — no straightening, no hassles. The Grippy Mat won’t budge while you work, making it easy to use a vacuum or water extractor for routine maintenance.
High-performance needlepunched fibers and low-profile surface design trap dirt, debris, sand and liquids to help prevent tracking; mat looks cleaner, longer.
The mat cuts easily without fraying or unraveling so you can create custom sizes and shapes; lets you work around beams, turn corners and cover asymmetrical floor layouts.
They are a cost-effective replacement for risky and outdated traditional rubber-backed entrance mats and scraper mats
Certifications: Tested and Certified by the National Floor Safety Institute (NFSI): when used as part of your floor safety program, high-traction surfaces such as PIG Grippy Mat have been proven to reduce slip-and-fall claims by as much as 90%.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty