Foxx Life Sciences PUREGRIP® HPLC Reservoir Caps and Bottle Assemblies offer the safest and most ergonomic lab safety solution in the industry. These assemblies come with VersaCap Technology, A Class VI PTFE Manifold and an Air Inlet Valve combined with a Foxx 25 mm, 0.2 µm PTFE Filter for safe vapor filtration.
The Air Inlet Valve and PTFE Filter block out all harmful solvent vapors while blocking any incoming contaminants from the incoming air. This valve threads directly into any of Foxx Life Sciences HPLC Reservoir Manifolds.
The sealed VersaCap design that Foxx offers eliminates tubes twisting as you thread the cap and eliminates the tubes from sliding in and out of the container. These HPLC Reservoir Assemblies guarantee you to get accurate data from your analysis with its air-tight seal using a Class VI PTFE Manifold and Chemically Resistant Viton O-Ring. Each assembly comes with fittings for both ¹/₈" and ¹/₁₆" OD Tubing along with corresponding plugs to fill any unused ports.