Would you like to log in again?
CONTAINER PG II SLIDING YLLOW 8 GAL CS5
Supplier: Cardinal Health
Container Pg Ii Sliding Yllow 8 Gal CS5^ Container, PG II Chemotherapy, Yellow, sliding Lid, Absorbent pad helps contain excess fluid, Label differentiates containers from those not D.O.T. Compliant, Molded-in carrying handles allow for easier transport, 17.75inH x 11inD x 15.5inW, 8 gal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONTAINER PG II SLIDING YLLOW 8 GAL CS5 76380-242 8985SPG2 Case of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty