Would you like to log in again?
CONTAINER PG II HINGED YELLOW 18GAL CS5
Supplier: Cardinal Health
Container Pg Ii Hinged Yellow 18Gal CS5^ Container, PG II Chemotherapy, Yellow, Hinged Lid, Absorbent pad helps contain excess fluid, Label differentiates containers from those not D.O.T. Compliant, Molded-in carrying handles allow for easier transport, 26inH x 12.75inD x 18.25inW, 18 gal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONTAINER PG II HINGED YELLOW 18GAL CS5 76380-246 8989PG2 Case of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty