Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Clorox® Hydrogen Peroxide Disinfecting Wipes, Janitorial Supplies
Supplier: Essendant
Clorox®
CLO30824 CLO30825 CLO30826 CLO30827
76380-494CT 224.92 USD InStock
76380-494 76380-492 76380-498 76380-496
Clorox® Hydrogen Peroxide Disinfecting Wipes, Janitorial Supplies
Wipes Disposable Wipes and Towels Disinfectant Wipes
Clorox® Hydrogen peroxide disinfecting wipes disinfect and clean surfaces quickly while killing germs and viruses in as little as 30 seconds.
  • Pre-moistened germicidal wipes
  • Active ingredient: Hydrogen peroxide
  • Use this one-step disinfecting cleaner on a broad range of surfaces
These wipes are non-corrosive and have no harsh fumes, bleach, VOCs, or added fragrances. Wipes are EPA registered to kill more than 40 microorganisms, including 13 antibiotic-resistant organisms. Use in Healthcare settings including hospitals, medical facilities, dental offices, veterinary offices, and long-term care facilities.
Caution: Please refer to the manufacturer's instructions for use for additional data regarding claims.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty