Would you like to log in again?
Strep-Tactin® XT Sepharose Chromatography Resins, Cytiva
Supplier: Cytiva
Sepharose™
29401324 29401326 29401326 29401324
76382-794EA 4735.57 USD InStock
CA76382-794 CA76382-792 76382-794 76382-792
Strep-Tactin® XT Sepharose Chromatography Resins, Cytiva
Protein Purification Protein Purification Resins
A chromatography resin for purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®. These proteins bind very specifically to the immobilized Strep-Tactin XT ligand giving highly pure target protein.
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration
  • Compatible with a wide range of detergents, denaturants, and other additives
Strep-Tactin XT sepharose contains the latest Strep-Tactin ligand, XT, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein than with the predecessor product, StrepTactin sepharose high performance (HP). Strep-Tactin XT sepharose also enables purification under denaturing conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty