Would you like to log in again?
Borosil® Low-Form Measuring Beakers
Supplier: Foxx Life Sciences
Borosil®
1000D33 1000D18 1000D25 1000D30 1000D38 1000D31 1000005 1000D12 1000009 1000D29 1000D24 1000D28 1000D16 1000006 1000D23 1000D21
76383-170EA 313.04 USD InStock
76383-170 76383-160 76383-150 76383-162 76383-134 76383-156 76383-168 76383-146 76383-158 76383-136 76383-148 76383-152 76383-164 76383-154 76383-166 76383-138
Borosil® Low-Form Measuring Beakers
Beakers
Ideal for general industry or education laboratory use and a wide variety of other applications. Borosil® low-form beakers are the ideal container for use when stirring, mixing, or heating liquids.
  • 3.3 Borosilicate glass chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
  • With embossed scale, white graduations (from size 25 ml on) and spout
  • Uniform wall thickness
  • Easy pour spout
Full range of beakers for every application.
Each beaker has uniform wall thickness distribution which aids in the balance between thermal resistance and mechanical strength, which is ideal for heating liquids. The pouring spout design allows for clean and precise pouring. Each beaker (except for sizes 5 ml and 10 ml) comes graduated to assist in measuring content. All sizes come standard with a durable enameled space for marking.
Certifications: Complies with ISO 2619, ISO 3819 and DIN 12331.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty