Would you like to log in again?
Borosil® Measuring Cylinders, Graduated, Pour-Out Single TC, Class B
Supplier: Foxx Life Sciences
Borosil®
3027021 3027006 3027005 3027009 3027024 3027012 3027030 3027016 3027029
76383-222CS 283.9 USD InStock
76383-222 76383-224 76383-230 76383-220 76383-232 76383-226 76383-216 76383-228 76383-218
Borosil® Measuring Cylinders, Graduated, Pour-Out Single TC, Class B
Cylinders Measuring Cylinders
Borosil® graduated measuring cylinder, pour out, single metric, Class B accuracy as per ASTM E 1272 with hexagonal base.
  • Calabrated per Class B tolerences
  • Graduation in amber enamel
  • 3.3 borosilicate glass
  • Sturdy hexagonal base
Borosil® measuring cylinders comply with ASTM E 1272. Calibrated as per Class B tolerances, each cylinder is engraved with a unique serial number and comes with a Certificate of Calibration traceable to national standards. The graduations are marked with amber enamel and the cylinder is manufactured with chemically resistant 3.3 borosilicate glass. Its sturdy hexagonal base prevents roll-over issues and ensures for better stability.
Certifications: In compliance with USP tolerance standards. ASTM 1272.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty