Would you like to log in again?
Avantor® ACE® C4-300, HPLC Columns, 10 µm
Supplier: VWR
ACE®
ACE-233-1520 ACE-233-0508 ACE-233-1220 ACE-233-0346 ACE-233-7520 ACE-233-0103GD ACE-233-1508 ACE-233-2508 ACE-233-0510 ACE-233-1246 ACE-233-1546 ACE-233-2510 ACE-233-0120GD ACE-233-0546 ACE-233-0520 ACE-233-7546 ACE-233-3546 ACE-233-2520 ACE-233-0110GD ACE-233-1510 ACE-233-2546 ACE-233-1046 ACE-233-0246 ACE-233-1010 ACE-233-1008 ACE-233-1020
76391-318EA 714.79 USD InStock
76391-318 76391-358 76391-314 76391-336 76391-316 76391-354 76391-332 76391-312 76391-356 76391-334 76391-350 76391-352 76391-330 76391-370 76391-328 76391-368 76391-346 76391-324 76391-326 76391-348 76391-342 76391-320 76391-322 76391-344 76391-360 76391-362
Avantor® ACE® C4-300, HPLC Columns, 10 µm
Chromatography Columns
Avantor® ACE® C4-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Designed for peptide and protein separations
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty