Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 3 µm

Supplier: VWR
Avantor® ACE® Excel®
EXL-1113-2503U EXL-1113-1002U EXL-1113-0346U EXL-1113-1246U EXL-1113-0503U EXL-1113-1202U EXL-1113-1003U EXL-1113-1502U EXL-1113-2046U EXL-1113-0203U EXL-1113-1503U EXL-1113-7502U EXL-1113-3502U EXL-1113-0546U EXL-1113-1203U EXL-1113-3546U EXL-1113-1046U EXL-1113-0246U EXL-1113-0202U EXL-1113-2502U EXL-1113-2546U EXL-1113-0303U EXL-1113-1546U EXL-1113-0502U EXL-1113-7546U EXL-1113-0302U EXL-1113-3503U EXL-1113-7503U
76393-304EA 1047.9 USD InStock
76393-304 76393-264 76393-286 76393-288 76393-266 76393-244 76393-268 76393-246 76393-300 76393-248 76393-302 76393-280 76393-260 76393-282 76393-284 76393-262 76393-274 76393-252 76393-254 76393-276 76393-278 76393-256 76393-258 76393-290 76393-270 76393-292 76393-272 76393-250
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.

  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases

These stainless steel columns are available in a wide range of dimensions from capillary to preparative for HPLC and UHPLC separations.

Ordering information: Method validation kits contain three different batches of the same phase - available on request.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty