Would you like to log in again?
VWR® 96-Well EVA and Silicone Microplate Covers
Supplier: Avantor
VWR®
VW60180-M105 VW60180-M178 VW08293813 VW60180-M123 VW60180-M116 VW60180-M113 VW08293640 VW60180-M121
76397-826PK 506.68 USD InStock
76397-826 76397-828 76397-820 76397-830 76397-822 76397-832 76397-824 76397-834
VWR® 96-Well EVA and Silicone Microplate Covers
Seals Microplate Seals
VWR® 96-well covers are designed for chromatography and high throughput screening (HTS) applications.
  • Covers for chromatography and storage
  • Ultra-pure silicone
  • SBS and ANSI standard footprint
  • Teflon barrier eliminates cross contamination of samples
  • 80 °C dry heat autoclavable
  • Resists coring and tearing
VWR® 96-well covers are manufactured specifically for chromatography. The multiple options ensure sample security, no matter the application. Non-cut and pre-cut versions cover the majority of all instrument requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty