Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottles, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nalgene®
CE-N2019-0125 CE-N2019-0030 CE-N2019-0250 CE-N2019-0500 CE-N2019-2000 CE-N2019-1000 CE-N2019-0060
76397-914CS 2022.43 USD InStock
76397-914 76397-916 76397-918 76397-870 76397-864 76397-866 76397-868
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottles, Thermo Scientific
Bottles Plastic Bottles
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.

  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Suitable for frozen storage down to –40 °C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Sterile and non-pyrogenic
  • Supported by extensive validation documentation

Bottles are washed with USP water for injection and handled post-washing in Class 5 cleanrooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than ¹/₃ the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10⁻⁶ inside the container/closure, including two heat-sealed sterile barriers to protect the product prior to use.

Packaging: Packed in shrink-wrapped trays. 2 L size features molded-in handgrips.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty