Would you like to log in again?
Nalgene® Biotainer Bottles, PETG, Clean, Certified, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
383110-42 383005-42 383415-16 383025-42 383230-42 383230-20 383415-42
76397-890CS 1195.39 USD InStock
76397-890 76397-892 76397-894 76397-886 76397-896 76397-888 76397-898
Nalgene® Biotainer Bottles, PETG, Clean, Certified, Thermo Scientific
Bottles Plastic Bottles
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Sterile
  • Clear finish
  • PETG construction
These bottles can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0.5 EU/ml endotoxin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty