Would you like to log in again?
Thermo Scientific™ Nalgene™ PC Certified Clean Biotainer Carboys, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
383030-42 383120-42 383233-42
76397-902CS 1186.89 USD InStock
76397-902 76397-904 76397-900
Thermo Scientific™ Nalgene™ PC Certified Clean Biotainer Carboys, Thermo Scientific
Bottles Carboys
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Sterile
  • Clear finish
  • Impact resistant polycarbonate construction
These carboys can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0.5 EU/ml endotoxin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty