Would you like to log in again?
Avantor® Hichrom, Chromatography Tubing, Precut SS
Supplier: VWR
10-SS-10 20-SS-10 05-SS-05 10-SS-30 10-SS-07 05-SS-10
76397-960PK 86.17 USD InStock
76397-960 76398-010 76397-962 76397-954 76397-964 76397-956
Avantor® Hichrom, Chromatography Tubing, Precut SS
Tubing Chromatography Tubing
316 Stainless steel, precision bore ¹/₁₆" type tubing.
  • Passivated for chemical resistance
  • Square, burr-free ends
  • Precut lengths
0.005 and 0.007" Int. Ø - suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
0.010" Int. Ø - suitable for connection of analytical and semi-preparative columns containing 5 and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.
0.20" Int. Ø - suitable for use in preparative systems or in non-critical connections (pump to sample injector where low flow resistance is more important than low internal volume).
Ext. Ø: 1.5 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty