Would you like to log in again?
Avantor® Hichrom, Chromatography Tubing, PEEK
Supplier: VWR
16-PEEK-05-5M 16-PEEK-20-5M 16-PEEK-20-10M 16-PEEK-03-10M 16-PEEK-05-10M 16-PEEK-07S-10 16-PEEK-07-5M 16-PEEK-30-10M 16-PEEK-20-25M 16-PEEK-04-10M 16-PEEK-07-10M 16-PEEK-30-5M 16-PEEK-10-10M 16-PEEK-03-5M 16-PEEK-07-3M 16-PEEK-10-5M 16-PEEK-02-10M
76397-990EA 253.37 USD InStock
76397-990 76397-992 76397-970 76397-980 76397-994 76397-972 76398-000 76397-982 76397-996 76397-974 76397-984 76397-976 76397-998 76397-986 76397-978 76397-988 76397-968
Avantor® Hichrom, Chromatography Tubing, PEEK
Tubing Chromatography Tubing
PEEK (polyetheretherketone) polymer tubing is biocompatible and chemically inert to most solvents. It is completely flexible, can easily be cut to desired lengths and is suitable for a wide range of applications.
  • Biocompatible, inert and easily cut
  • Good for high pressure applications
  • Can be used with stainless steel or polymer fittings
  • ¹/₁₆" O.D. tubing
It has a very smooth internal surface and is the least permeable to gas of all the polymer tubing materials. Alternative O.D. tubing, example ¹/₈" for high pressure semi-preparative systems and 360 μm for capillary systems, is also available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty