Would you like to log in again?
Avantor® Hichrom, HPLC Column Connectors
Supplier: VWR
Avantor® Hichrom
HI-1091
76399-058EA 51.2 USD InStock
76399-058
Avantor® Hichrom, HPLC Column Connectors
Fittings and Connectors
These column connectors are used to connect Hichrom guard and analytical columns.
  • Stainless steel connector
  • Female-female end fittings
The connector consists of a stainless steel tubing with appropriate 1/16" nuts and ferrules and can be used to connect female end fitting Hichrom guard and analytical columns and make similar connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty