Would you like to log in again?
Avantor® Hichrom, Static Mixing Tee
Supplier: VWR
Avantor® Hichrom
HI-454
76399-366EA 227.5 USD InStock
76399-366
Avantor® Hichrom, Static Mixing Tee
Fittings and Connectors
These static mixing tee are ideal for microbore or analytical gradient HPLC. It has a low swept volume (2.2 µl) and is designed for flow rates of 0.5 to 3 ml/min.
  • PEEK body with two-piece fingertight fittings
  • Low swept volume
  • Maximum pressure of 5000 psi
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty