Would you like to log in again?
Avantor® Partisil Silica, HPLC Columns
Supplier: VWR
Partisil®
P5-125A P10-10C5 P10-150A P5-250A P5-250AF P10-150AF P10-50A P5-150AF P5-250P P5-50A P10-150AS P10-100A P5-10CM5 P5-100A P10-50AM P10-250AS P5-100AF P5-150A P10-50AS P5-50AS P10-250A P10-50AF P10-150AM P10-10CM5 P10-300AF P10-250AM P5-150AM P5-250AS P5-250AM P10-200A P5-50AM P5-10C5 P5-100AM P5-50AF P10-100AM P5-150AS P5-200A P10-100AF P10-100AS
76402-050EA 1791.58 USD InStock
76402-050 76401-706 76401-728 76402-052 76402-030 76401-704 76401-726 76401-724 76401-702 76401-700 76401-722 76402-036 76402-058 76402-038 76402-032 76402-054 76402-034 76402-056 76401-708 76401-690 76402-028 76401-710 76401-698 76401-694 76401-692 76401-718 76402-060 76401-716 76402-040 76401-714 76401-712 76402-046 76402-024 76402-048 76402-026 76402-042 76402-044 76402-022 76401-720
Avantor® Partisil Silica, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas.
  • Irregular porous silica
  • 5 and 10 µm particle sizes
  • Hichrom high efficiency
A large surface area gives it a high loading capacity. Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty