Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SP Bel-Art Sterileware® Pharma Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art
Sterileware®
F36909-0125 F36909-0250 F36910-1000 F36910-0030 F36909-0060 F36910-2500 F36910-0060 F36909-0500 F36909-0030 F36909-1000 F36910-0125 F36910-0250 F36910-0500 F36909-2500
76402-872CS 1015.94 USD InStock
76402-872 76402-860 76402-862 76402-870 76469-934 76469-944 76402-868 76469-936 76469-946 76402-864 76469-940 76402-866 76469-942 76469-938
SP Bel-Art Sterileware® Pharma Scoops, Bel-Art Products, a part of SP
Scoops
Ideal for sampling for both production work and laboratory use, Pharma Scoops are used by pharmaceutical, food, cosmetic, and chemical companies.
  • Bright red color reduces chance of scoop being left behind in a sample; can serve as code to avoid cross contamination; can provide contrasting background for light colored samples
  • High handle location keeps hand further from sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty