Would you like to log in again?
VIAL CRYO.S PP CONICAL BOTTOM 1ML PK10
Supplier: Greiner Bio-One
Vial Cryo.S Pp Conical Bottom 1Ml PK10^ cryo.s* Vial, Material: polypropylene, conical bottom, internal thread, natural screw cap, writing area, starfoot, natural, sterile, Size: 1 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL CRYO.S PP CONICAL BOTTOM 1ML PK10 76402-924 123263-TRI Case of 200 Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty