Would you like to log in again?
Corepoint Scientific™ General Purpose Flammable Storage Freezers, Horizon Scientific
Supplier: Horizon Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corepoint Scientific™
FF301WWW/0MGP
76402-978EA 10357.9 USD InStock
76402-978
Corepoint Scientific™ General Purpose Flammable Storage Freezers, Horizon Scientific
Freezers Flammable Material Freezers
These flammable storage freezers are designed to store and protect flammable substances in laboratory settings.
  • Mechanical thermostat
  • Interior shelving
  • Keyed door lock
  • Probe access
  • Manual defrost
  • Meet NFPA guidelines
These freezers are designed to store and protect volatile substances in laboratory settings. These units meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers.
Ordering information: All units come with a one year parts and labor warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty