Would you like to log in again?
First Aid Only™ Standard Pro Bleeding Control Kit, Acme United
Supplier: Acme United
First Aid Only™
91136-002
76403-162EA 122.7 USD InStock
76403-162
First Aid Only™ Standard Pro Bleeding Control Kit, Acme United
First Aid Supplies First Aid Kits
Bleeding control kits empower a first responder bystander with the right tools to stop life-threatening blood loss before professional help arrives.
  • Handle most multiple-wound limb or torso bleeding wounds
  • Windlass-style and a back-up limb tourniquet
  • Hemostatic blood-clotting granule packet
  • Highly absorbent Israeli bandage with a pressure applicator
  • Aluminized blanket to keep the victim warm
The standard pro bleeding control kit provides all the tools required to handle most limb or torso bleeding wounds. It is equipped with a marker to denote the time the tourniquet was applied.
Packaging: The kit comes in an organized, easy access pouch, allowing users to find all necessary supplies in a hurry.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty