Would you like to log in again?
Nalgene™ Non-Phthalate PVC Tubing, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
8701-9030 8701-0030 8701-9090 8701-4060 8701-0310 8701-9020 8701-0080 8701-4140 8701-0190 8701-0090 8701-0350 8701-4120 8701-0060 8701-0120 8701-0140 8701-0160 8701-0110 8701-0180 8701-0010 8701-0260 8701-0020 8701-0070 8701-0100 8701-0200 8701-0130
76403-558CS 302.74 USD InStock
76403-558 76403-578 76403-556 76403-582 76403-560 76403-580 76403-542 76403-586 76403-564 76403-562 76403-584 76403-546 76403-568 76403-588 76403-566 76403-544 76403-548 76403-590 76403-550 76403-572 76403-570 76403-554 76403-576 76403-552 76403-574
Nalgene™ Non-Phthalate PVC Tubing, Thermo Scientific
Tubing Transfer Tubing
Nalgene Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticizers.
  • Compare to Tygon® E-3603 tubing
  • Marked with Int. Ø and regulatory compliance as well as at 12" intervals for easy identification and measurement
  • Durometer hardness is Shore A, 60
Nalgene™ Non-Phthalate PVC Tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene 180 tubing but is free from orthophthalate DEHP plasticizers. Resists hardening and discoloration from a wide range of laboratory chemicals (demonstrates dimensional stability) and does not impart odor or taste making this tubing ideal for a wide range of non-medial, general lab, research and pharmaceutical applications.
Certifications: Meets Food Additives Amendment of the Federal Food, Drug, and Cosmetic Act. Meets the requirements of the U.S. Pharmacopoeia Class VI Biological Tests for Plastics, USP <88>. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and California Proposition 65 standards
Caution: Not intended for medical use.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty