Would you like to log in again?
VWR TUBING PVC 1/4X3/8X1/16IN 50FT CS1
Supplier: VWR International
Vwr Tubing Pvc 1/4X3/8X1/16In 50Ft CS1^ VWR Tubing, Clear, made by highest grade resins, Non-Phthalate, Material: PVC, Resists a wide range of chemicals, Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, Autoclavable, Dimensions: 1/4X3/8X1/16in, Length: 50FT
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR TUBING PVC 1/4X3/8X1/16IN 50FT CS1 76403-632 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty