Would you like to log in again?
METAL COMBINATION ELECTRODE AGCL 62
Supplier: XYLEM/YSI
Metal Combination Electrode Agcl 62^ Metal combination electrode for halogenide titrations, Glass shaft, platinum junction, electrolyte KNO3 2 mol/l + KCl 0.001 mol/l, Ag/AgCl-reference system, sensor silver cap 4 mm dia, chlorinated, plug head, length 120 mm, 12 mm dia, -5 to +100 deg C
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
METAL COMBINATION ELECTRODE AGCL 62 76403-780 285102413 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty