Would you like to log in again?
PH COMBINATION ELECTRODE N62 120 MM LEN
Supplier: XYLEM/YSI
Ph Combination Electrode N62 120 Mm Len^ combination electrode, pH, N 62, with plug head for laboratory application, Glass shaft, platinum junction, electrolyte KCl 3 mol/l, Silamid reference system, sphere membrane, A-glass, -5.+100 degree C, pH: 0.14, length: 120 mm, Diameter: 12 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PH COMBINATION ELECTRODE N62 120 MM LEN 76403-802 285100034 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty