Would you like to log in again?
AlphaTec® 37-300 Light-Duty Nitrile Gloves, Ansell
Supplier: Ansell
AlphaTec®
37300110 37300100 37300090 837155 37300070 37300090 37300110 37300070 37300080 37300080
76404-190PK 23.46 USD InStock
CA76404-190 76404-190 76404-192 76404-184 CA76404-184 CA76404-192 76404-186 76404-188 CA76404-188 CA76404-186
AlphaTec® 37-300 Light-Duty Nitrile Gloves, Ansell
Gloves
All day comfort at exceptional value.
  • 8 mil/0.2 mm thickness allows dexterity and tactility without a loss of durability
  • NBR offers protection against light acids, greases, oils, and liquids
  • Engineered fit allows optimum balance of fit and flexibility, use alone or as an over-glove
  • Latex-free formulation presents no risk of Type 1 allergies
  • Long cuff extends to the forearm
High-quality Nitrile gloves allow dexterity and tactililty while providing protection against light acids, oils, and liquids. Recommended for all food processing applications that involve food contact, including fruit processing, poultry processing, meal preparation, and dairy processing. Engineered with a long cuff that extends to the forearm, the 37-300 series of gloves maintain durability and flexibility on their own or as an over-glove.
Certifications: EN ISO 374 - S:2016; EN ISO 374-1:2016 Type B JKT; EN 388:2016 2001X; EN 420:2003 + A1:2009; EN 421; ISO 18889 G1
Packaging: Gloves are packaged 1 pair per inner bag, 12 inner bags per master bag, 12 master bags per carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty