Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Eppendorf® Barcoded CryoStorage Vials
Supplier: Eppendorf
0030079450 0030079400 0030079493 0030079485 0030079442 0030079477 0030079418 0030079469 0030079426 0030079434
76404-522EA 904.78 USD InStock
76404-522 76404-510 76404-524 76404-512 76404-526 76404-514 76404-516 76404-508 76404-518 76404-520
Eppendorf® Barcoded CryoStorage Vials
Vials
Eppendorf CryoStorage vials provide easy and fast sample recognition for safe sample identification through three-level coding: 1D barcode (side), Clear coding (side), and 2D datamatrix code with ECC 200 error correction (bottom).
  • ECC 200 error correction function enables safe code reading even when codes are up to 30% damaged
  • Temperature range: −196 (gas phase of LN₂) to +121 °C
  • Manufactured from high-quality polypropylene in class VIII cleanroom for highest sample purity
  • Pre-capped and pre-racked for comfortable handling
  • Direct import into Eppendorf eLABInventory and eLABJournal software for convenient handling
These Vials have an automation-compatible lid style. The inner shape of the vial lid fits to standard de-capper systems. The twistlock bottom of the vial prevents it from rotating within the rack when being opened.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty