Would you like to log in again?
Corning® LSE™ Mini Incubator, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning® LSE
6800
76404-716CS 536.77 USD InStock
76404-716
Corning® LSE™ Mini Incubator, Corning
Incubators Compact Benchtop Incubators
The compact mini incubator is designed for personal use, it is ideal for small laboratories and classrooms.
A thermometer port in the top of the unit allows the user to monitor temperatures. The large window in the door gives a clear view of the contents.
Certifications: cETLus
Ordering information: Supplied with one shelf. Addditional spare shelves and safety thermometer are available as optional accessories.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty