Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clean and Sweep Store-Boards™ Kits, Accuform®

Supplier: Accuform

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PSB716WTBK PSB116WTBK PSB116YLBK PSB716GNWT PSB616RDWT PSB116RDWT PSB616YLBK PSB116BUWT PSB716RDBK PSB616BUWT PSB716GNBK PSB716BUWT PSB616BUBK PSB616GNWT PSB116GNWT PSB616GNBK PSB116WTWT PSB716YLWT PSB116GNBK PSB716RDWT PSB716YLBK PSB616YLWT PSB716WTWT PSB616WTBK PSB116RDBK PSB616RDBK PSB116BUBK PSB116YLWT PSB716BUBK PSB616WTWT
76404-780KT 620.42 USD InStock
76404-780 76404-784 76404-762 76404-740 76404-782 76404-760 76404-776 76404-732 76404-754 76404-752 76404-774 76404-736 76404-758 76404-756 76404-778 76404-734 76404-738 76404-790 76404-750 76404-772 76404-770 76404-766 76404-744 76404-788 76404-742 76404-764 76404-786 76404-748 76404-746 76404-768
Clean and Sweep Store-Boards™ Kits, Accuform®
Boxes Storage Boxes
Clean and Sweep Store-Boards™ Kit keeps tools organized.

  • Promote the concept, a place for everything and everything in its place
  • At a glance, see that tools and supplies are in their designated storage location
  • Tools included
  • Material options: Accu-Shield™, Ultra Aluma-Lite™ or Max-Duty Aluminum
  • Board measurement: 172.72H×76.2W cm

Accu-Shield™ - Highest performance material, is a two-ply, polycarbonate plastic. Clear, non-glare front provides heavy duty protection against chemicals, corrosion and abrasions. Subsurface, UV-printed image, encapsulated on the back by poly laminate, will continue to look good for years while maintaining superior strength. Service temperature: –60 to 260 °F.

Ultra Aluma-Lite™ - Composite plastic bonded with two layers of aluminum making it light-weight, 40% lighter than solid aluminum, but extremely durable. Corrosion-, impact-, and UV-resistant making it an ideal choice for indoor or outdoor use. Service temperature: –40 to 170 °F.

Max-Duty Aluminum - Provides the strength of metal yet never rusts. Finishing includes UV protective coating to prevent fading, chemical damage, and abrasions. Service temperature: -40° to 250 °F.

Delivery information: Kit includes: Store-Board™ (1), HRM195 Metal Dust Pan (1), HRM146 Lobby Dust Pan (1), HRM147 60" Push Broom Handle (1), HRM125 24" Push Broom Head (1), HRM126 Brush (1), HRM130 Angle Broom (1) and zinc-plated hardware for hanging tools onto board.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty