Would you like to log in again?
Dry Zone Store-Boards™ Kits, Accuform®
Supplier: Accuform
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PSB712GNBK PSB712GTWT PSB612BURD PSB712GNRD PSB612BUGN PSB612YLBK PSB712BURD PSB712BUYL PSB712GTBK PSB712WTBK PSB712WTGN PSB712BUBU PSB712RDBK PSB712RDRD PSB712GNWT PSB612GNBU PSB612YLBU PSB612RDBK PSB612WTRD PSB612GNYL PSB712GNBU PSB712WTYL PSB612WTBK PSB712WTRD PSB712BUGN PSB712WTBU PSB712WTWT PSB612YLGN PSB612YLRD PSB612GNBK PSB612YLYL PSB712BUBK PSB612GTYL PSB612GTBU PSB612WTBU PSB712RDWT PSB712GNGN PSB712GTRD PSB612GNRD PSB612GTRD PSB612WTGN PSB612YLWT PSB712GTBU PSB612BUYL PSB612BUWT PSB712YLRD PSB612BUBU PSB712RDYL PSB612RDRD PSB712RDBU PSB712YLBU PSB712YLGN PSB612GTBK PSB612RDBU PSB612RDGN PSB712GNYL PSB612WTWT PSB612GNWT PSB712BUWT PSB712GTGN PSB612GNGN PSB712GTYL PSB612WTYL PSB612GTWT PSB712YLWT PSB612BUBK PSB712RDGN PSB612RDYL PSB712YLYL PSB712YLBK PSB612GTGN PSB612RDWT
76405-300KT 1062.4 USD InStock
76405-300 76405-322 76405-344 76405-302 76405-346 76405-324 76405-326 76405-304 76405-348 76405-328 76405-306 76405-308 76405-290 76405-292 76405-294 76405-350 76405-296 76405-330 76405-352 76405-298 76405-332 76405-310 76405-354 76405-334 76405-312 76405-336 76405-314 76405-316 76405-338 76405-318 76405-284 76405-340 76405-286 76405-320 76405-342 76405-288
Dry Zone Store-Boards™ Kits, Accuform®
Boxes Storage Boxes
Promote cleanliness and safety by utilizing Store-Boards™ in a HACCP Management System Dry Zone Critical Control Point. Prevent cross contamination by keeping specified equipment separate and easily identified.
  • Promote the concept, a place for everything and everything in its place!
  • At a glance, see that tools and supplies are in their designated storage location
  • Tools included
  • Board measures 68H×30"W
Material Options:
Ultra Aluma-Lite™ - Composite plastic bonded with two layers of aluminum making it light-weight, 40% lighter than solid aluminum, but extremely durable. Corrosion-, impact-, and UV-resistant making it an ideal choice for indoor or outdoor use. Service temperature: −40 to 170 °F.
Max-Duty Aluminum - Provides the strength of metal yet never rusts. Finishing includes UV protective coating to prevent fading, chemical damage, and abrasions. Service temperature: −40 to 250 °F.
Delivery information: Kit includes: (1) Store-Board™, (1) HRM121 Vikan Soft Baker’s Brush, (1) HRM123 Vikan Angle Broom, (1) HRM142 Vikan 51" Plastic Handle, (1) HRM127 Remco Flat Broom, (1) HRM140 Vikan 59" Plastic Handle, (1) HRM196 Vikan Dust Pan, (1) HRM160 Remco 14" Shovel, and stainless steel hardware for hanging tools onto board.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty