Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ABC Fruit Blend (Apple, Blueberry, Coconut)

Supplier: Bio-Serv
F7228 F7228 S7228 S7228
76406-470BG 32.24 USD InStock
CA76406-470 76406-470 CA76406-498 76406-498
ABC Fruit Blend (Apple, Blueberry, Coconut)
Animal Supplies Animal Feed
A naturally sweet multi-species treat. Delicious foraging mixture of dried apples, dried blueberries and coconut.

  • Protein: 4.0%
  • Fat: 28.5%
  • Fiber: 8.0%
  • Ash: 1.5%
  • Moisture: <12%
  • Carbohydrate: 48.0%
  • Calories: 4.64 kcal/gm

Ingredients: Diced apple, coconut chips/flakes, blueberries

Certifications: Certified (contaminant screened).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty