Would you like to log in again?
Rat MD's™ Baytril Bacon Flavor
Supplier: Bio-Serv
Rat MD's™
MD300-2 MD300-2
76406-482PK 121.01 USD InStock
CA76406-482 76406-482
Rat MD's™ Baytril Bacon Flavor
Animal Supplies Animal Feed
Baytril is an effective antibiotic drug used in a variety of species including rodents.
Baytril MD's are highly palatable and encourage appetite post-operatively promoting a more rapid recovery versus other conventional methods. They are an ideal pain-free alternative to stressful and injurious post-operative injections.
Baytril MD's can be customized to include anti-inflammatory drugs such as Rimadyl and Meloxicam to provide both medications easily and affordably.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty