Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rodent Foraging Crumbles, Certified, Bio-Serv

Supplier: Bio-Serv
F5783 S5783 S5783 F5783
76406-496BG 51.83 USD InStock
76406-496 CA71002-336 89408-276 CA76406-496
Rodent Foraging Crumbles, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete, grain-based treat

  • The tiny crumbles encourage foraging behavior
  • Scatter a pinch directly into bedding
  • Bacon flavor enhances foraging activity
  • Nutritionally assayed

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 3.41 kcal/g
Nutritional ash content: 7.4%
Nutritional carbohydrate content: 54%
Nutritional fat content: 4.3%
Nutritional fibre content: 4%
Nutritional moisture content: <10%
Nutritional protein content: 21.6%

Certifications: Certified (Contaminant Screened).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty