Would you like to log in again?
EARMUFF VS130M HIGH DIELECTRIC MULTI
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Earmuff Vs130M High Dielectric Multi^ VeriShield* Earmuff, VS130M dielectric multi-position earmuff, Comfortably fits a wide range of head sizes, Wide cup opening is ideal for workers with large ears, or who wear hearing aids, Memory foam ear cushions, NRR 30
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EARMUFF VS130M HIGH DIELECTRIC MULTI 76406-912 1035187-VS Case of 10 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty