Would you like to log in again?
GLOVES NITRILE BL 40MIL 12IN 8/M DZ12
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Gloves Nitrile Bl 40Mil 12In 8/M DZ12^ Nitri-Knit* Supported Nitrile Gloves, NK803, offer superior resistance to cuts, snags, abrasions and punctures, high resistance to solvents caustics and animal fats, Interlock knit liner, Thickness: 40 mil, Length: 12 in, Color: Blue, Size: 8, Medium
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES NITRILE BL 40MIL 12IN 8/M DZ12 76406-932 NK803/8-H5 Case of 144 Retrieving
Dozen Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty