Would you like to log in again?
ESD Brushes, Menda
Supplier: Menda
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35694 35688 35692 35691 35693 35687
76407-240EA 26.42 USD InStock
76407-240 76407-236 76407-238 76407-232 76407-244 76407-234
ESD Brushes, Menda
Brushes
ESD brushes are designed to help clean static sensitive equipment and components.
  • Ideal for removing dust, dirt, and foreign object debris from static sensitive components
  • Yellow bristles indicate dissipative brushes
  • Black bristles indicate conductive brushes
These ESD Brushes are designed to allow thorough cleaning of ESD sensitive assemblies without the danger of high charges generated by conventional brushes. Brushes are available with wither dissipative or conductive bristles.
Yellow dissipative bristles are highly resilient and abrasion resistant. They are hot stamped with the ESD protective symbol to indicate that they can be used in static sensitive applications. Black conductive bristles minimize static charge generation and remove electrostatic charges to ground. Conductive bristles are available in two styles, semi-fine and firm. Semi-fine bristles are ideal for chemical and electronics applications. Firm bristles are mainly for use with electronics, such as circuit boards.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty