Would you like to log in again?
D-Sub Conductive Connector Cover, Menda
Supplier: Menda
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35784
76407-246CS 992.42 USD InStock
76407-246
D-Sub Conductive Connector Cover, Menda
Covers
A cap to cover electrictical connectors.
  • Helps elimate dust and other particles in the connector
  • Made of conductive material
  • Ideal for clean applications
  • Free of carbon particulates; will not leave black marks
  • Noncorrosive
  • Contacts: 37
  • Connector type: Pin
Permanently volume conductive and free of carbon black particulate. The connector covers do not leave black marks and are more suited to clean situations. Meets the requirements of NASM5501 31/32. Connector covers are noncorrosive and produce no visible corrosion when tested to Federal Test Method 101C using Kovar test plates. The material used in the covers was selected for its resilience. Properly sized, the covers provide a snug, secure fit. This helps keep connectors clean and prevents the cover from accidentally falling off.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty